3,5-Dimethoxyphenyl isocyanate

Supplier: Matrix Scientific

030164-500MG
101780-498EA 146.15 USD
101780-498
3,5-Dimethoxyphenyl isocyanate
3,5-Dimethoxyphenyl isocyanate

Hazard: Toxic/Irritant

Formula: C₉H₉NO₃
MW: 179.18 g/mol
CAS Number: 54132-76-2
UN: 3335
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR