3,4,5-Trifluorobenzenesulfonyl chloride ≥98%
Supplier: Matrix Scientific
Synonyms:
3,4,5-Trifluorobenzene-1-sulfonyl chloride
, 3,4,5-Trifluorobenzene-1-sulphonyl chloride
026758-25G
101780-490EA
136.92
USD
101780-490
3,4,5-Trifluorobenzenesulfonyl chloride ≥98%
3,4,5-Trifluorobenzenesulphonyl chloride
Hazard: Flammable/Corrosive
Formula:
C₆H₂ClF₃O₂S MW: 230.59 g/mol Boiling Pt: 76…77 °C (760 mmHg) Hg |
MDL Number:
MFCD03094559 CAS Number: 351003-43-5 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 76-77°/760 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...