3,5-Dichloro-4-hydroxybenzenesulfonyl chloride ≥97%
Supplier: Matrix Scientific
024836-1G
024836-5G
101780-450EA
32.31
USD
101780-450
101780-452
3,5-Dichloro-4-hydroxybenzenesulfonyl chloride ≥97%
3,5-Dichloro-4-hydroxybenzenesulphonyl chloride
Hazard: Corrosive
Formula:
C₆H₃Cl₃O₃S Melting Pt: 123…130 °C |
MDL Number:
MFCD00171551 CAS Number: 13432-81-0 UN: 3261 ADR: 8,I |
Specification Test Results
Melting point | 118-119° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...