4-Acetylbenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Acetylbenzenesulfonyl Chloride4-Acetylbenzenesulphonyl Chloride4-Acetylbenzene-1-sulfonyl chloride4-Acetylbenzene-1-sulphonyl chloride

024828-5G
101780-448EA 83.08 USD
101780-448
4-Acetylbenzenesulfonyl chloride ≥97%
4-Acetylbenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₈H₇ClO₃S
MW: 218.66 g/mol
Melting Pt: 84…87 °C
MDL Number: MFCD00800269
CAS Number: 1788-10-9
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Melting point 84°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR