2-Fluoro-m-toluonitrile ≥98%
Supplier: Matrix Scientific
Synonyms:
2-Fluoro-3-methylbenzonitrile
024822-5G
024822-25G
101780-444EA
90.77
USD
101780-444
101780-442
2-Fluoro-m-toluonitrile ≥98%
2-Fluoro-m-toluonitrile
Hazard: Toxic
Formula:
C₈H₆FN MW: 135.1384 g/mol |
MDL Number:
MFCD07782078 CAS Number: 185147-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...