2,2,6,6-Tetramethylpiperidine ≥98%

Supplier: Matrix Scientific

024756-100G 024756-1KG 024756-500G
102551-070EA 241.54 USD
102551-070 101780-430 102551-072
2,2,6,6-Tetramethylpiperidine ≥98%
2,2,6,6-Tetramethylpiperidine

Hazard: Flammable

Formula: C₉H₁₉N
MW: 141.26 g/mol
Boiling Pt: 152 °C
Density: 0.837
MDL Number: MFCD00005985
CAS Number: 768-66-1

Order Now

Specification Test Results

Boiling point 152°
Density 0.837
ND20 1.445

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR