2-Chloro-6-methoxyisonicotinoyl chloride ≥95%

Supplier: Matrix Scientific

024694-1G
101780-416EA 115.38 USD
101780-416
2-Chloro-6-methoxyisonicotinoyl chloride ≥95%
2-Chloro-6-methoxyisonicotinoyl chloride

Hazard: Corrosive/Irritant/Moisture Sensitive

Formula: C₇H₅Cl₂NO₂
Boiling Pt: 79 °C (0.26 mmHg)
MDL Number: MFCD00125355
CAS Number: 116853-97-5

Order Now

Specification Test Results

Boiling point 79°/0.26mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR