3-Bromo-2,6-dichloropyridine ≥98%
Supplier: Matrix Scientific
024646-5G
101780-402EA
56.92
USD
101780-402
3-Bromo-2,6-dichloropyridine ≥98%
3-Bromo-2,6-dichloropyridine
Hazard: Toxic
Formula:
C₅H₂BrCl₂N Melting Pt: 68…71 °C |
MDL Number:
MFCD06798231 CAS Number: 866755-20-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...