2-Fluoro-4-iodobenzonitrile ≥98%

Supplier: Matrix Scientific

024633-25G 024633-5G
101780-396EA 292.31 USD
101780-396 101780-398
2-Fluoro-4-iodobenzonitrile ≥98%
2-Fluoro-4-iodobenzonitrile

Hazard: Toxic/Irritant/Irritant-Harmful

Formula: C₇H₃FIN
MW: 247.01 g/mol
MDL Number: MFCD03094172
CAS Number: 137553-42-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR