2-Methylcyclopentanone ≥96%

Supplier: Matrix Scientific

024596-25G 024596-100G 024596-5G
102551-062EA 214.46 USD
102551-062 102551-064 101780-374
2-Methylcyclopentanone ≥96%
2-Methylcyclopentanone

Purity: 96%
CAS: 1120-72-5
MDL Number: MFCD00001414
Molecular Formula: C6H10O
Molecular Weight: 98,15
Boiling Point: 139°C
Hazard: Flammable

Formula: C₆H₁₀O
MW: 98,14 g/mol
Boiling Pt: 139°C
Melting Pt: -75 °C
Density: 0,917
Flash Pt: 26°C
MDL Number: MFCD00001414
CAS Number: 1120-72-5
EINECS: 214-318-4
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Boiling point 139°
Density 0.917

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR