Trimethylsilyl cyanide ≥97%
Supplier: Matrix Scientific
023806-5G
023806-25G
023806-100G
101780-346EA
250.77
USD
101780-346
101780-342
101780-344
Trimethylsilyl cyanide ≥97%
Trimethylsilyl cyanide
Hazard: Flammable/Toxic by inhalation/Refrigerate
Formula:
C₄H₉NSi MW: 99.21 g/mol Boiling Pt: 118…119 °C Melting Pt: 11…12 °C Density: 0.744 |
MDL Number:
MFCD00001765 CAS Number: 7677-24-9 UN: 3383 ADR: 6.1,I |
Specification Test Results
Boiling point | 118-119° |
Density | 0.744 |
ND20 | 1.392 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...