4-Fluoro-m-toluonitrile ≥97%

Supplier: Matrix Scientific

021120-5G 021120-25G
101780-286EA 16.92 USD
101780-286 101780-288
4-Fluoro-m-toluonitrile ≥97%
4-Fluoro-m-toluonitrile

Hazard: Toxic

Formula: C₈H₆FN
MW: 135.14 g/mol
MDL Number: MFCD03788510
CAS Number: 185147-08-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR