4-Fluoro-2-methylbenzenesulfonyl chloride ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Fluoro-2-methylbenzene-1-sulfonyl chloride
, 4-Fluoro-2-methylbenzene-1-sulphonyl chloride
021068-1G
021068-5G
101780-276EA
18.46
USD
101780-276
101780-278
4-Fluoro-2-methylbenzenesulfonyl chloride ≥97%
4-Fluoro-2-methylbenzenesulphonyl chloride
Hazard: Corrosive
Formula:
C₇H₆ClFO₂S MW: 208.64 g/mol Boiling Pt: 245…246 °C Density: 1.433 |
MDL Number:
MFCD03094390 CAS Number: 7079-48-3 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 245-246° |
Density | 1.433 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...