2-Chloro-6-methylbenzaldehyde
Supplier: Matrix Scientific
020177-1G
020177-5G
101780-242EA
80
USD
101780-242
101780-244
2-Chloro-6-methylbenzaldehyde
2-Chloro-6-methylbenzaldehyde
Hazard: Toxic/Irritant
Formula:
C₈H₇ClO Boiling Pt: 74 °C (0.4 mmHg) Melting Pt: 39…40 °C |
MDL Number:
MFCD01934409 CAS Number: 1194-64-5 |
Specification Test Results
Boiling point | 74°/0.4mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...