(2R)-2-(Trifluoromethyl)pyrrolidine

Supplier: Matrix Scientific

Synonyms: 2(R)-2-Trifluoromethylpyrrolidine

018286-1G
101780-218EA 58.46 USD
101780-218
(2R)-2-(Trifluoromethyl)pyrrolidine
(2R)-2-(Trifluoromethyl)pyrrolidine

Hazard: Corrosive/Store in Freezer

Formula: C₅H₈F₃N
MW: 139.119 g/mol
Boiling Pt: 110…111 °C
Melting Pt: 27…31 °C
MDL Number: MFCD07784180
CAS Number: 1073556-31-6

Order Now

Specification Test Results

Boiling point 110-111°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR