3,5-Dimethylbenzenesulfonyl chloride ≥97%
Supplier: Matrix Scientific
Synonyms:
3,5-Dimethylbenzene-1-sulfonyl chloride
, m-Xylene-5-sulfonyl chloride
, 3,5-Dimethylbenzene-1-sulphonyl chloride
, m-Xylene-5-sulphonyl chloride
018272-25G
018272-1G
018272-5G
101780-214EA
44.62
USD
101780-214
101780-216
101780-212
3,5-Dimethylbenzenesulfonyl chloride ≥97%
3,5-Dimethylbenzenesulphonyl chloride
Hazard: Corrosive/Irritant/Refrigerate
Formula:
C₈H₉ClO₂S MW: 204.68 g/mol Melting Pt: 90…94 °C |
MDL Number:
MFCD03094655 CAS Number: 2905-27-3 UN: 3261 ADR: 8,II |
Specification Test Results
Melting point | 90-94° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...