2-Trifluoromethyl acrylonitrile ≥97%

Supplier: Matrix Scientific

012895-5G
101780-164EA 1130.77 USD
101780-164
2-Trifluoromethyl acrylonitrile ≥97%
2-Trifluoromethyl acrylonitrile

Hazard: Corrosive/Irritant

Formula: C₄H₂F₃N
MW: 121.06 g/mol
Boiling Pt: 76…77 °C
Density: 1.1753 (26 °C)
MDL Number: MFCD01673877
CAS Number: 381-84-0

Order Now

Specification Test Results

Boiling point 76-77°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR