4-Bromotetrafluorophenol ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2,3,5,6-tetrafluorophenol

012838-5G
101780-154EA 150.77 USD
101780-154
4-Bromotetrafluorophenol ≥97%
4-Bromotetrafluorophenol

Hazard: Corrosive/Irritant

Formula: C₆HBrF₄O
MW: 244.98 g/mol
Boiling Pt: 76…77 °C (4 mmHg)
Melting Pt: 28…30 °C
MDL Number: MFCD06246024
CAS Number: 1998-61-4

Order Now

Specification Test Results

Boiling point 76-77°/4mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR