2-Fluoro-5-trifluoromethylbenzyl chloride ≥98%

Supplier: Matrix Scientific

012618-5G 012618-25G
101780-142EA 21.54 USD
101780-142 101780-144
2-Fluoro-5-trifluoromethylbenzyl chloride ≥98%
2-Fluoro-5-trifluoromethylbenzyl chloride

Hazard: Corrosive/Irritant/Lachrymator

Formula: C₈H₅ClF₄
MW: 212.58 g/mol
MDL Number: MFCD05864315
CAS Number: 883543-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR