4-Nitro-2-(trifluoromethyl)benzonitrile ≥98%
Supplier: Matrix Scientific
012617-5G
012617-25G
101780-140EA
213.85
USD
101780-140
101780-138
4-Nitro-2-(trifluoromethyl)benzonitrile ≥98%
4-Nitro-2-(trifluoromethyl)benzonitrile
Hazard: Toxic/Irritant
Formula:
C₈H₃F₃N₂O₂ Melting Pt: 52…54 °C |
MDL Number:
MFCD01075729 CAS Number: 320-47-8 UN: 3439 ADR: 6.1,III |
Specification Test Results
Melting point | 52-54° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...