2-(Tributylstannyl)benzofuran

Supplier: Matrix Scientific

011160-1G
101780-048EA 653.85 USD
101780-048
2-(Tributylstannyl)benzofuran
2-(Tributylstannyl)benzofuran

Hazard: Irritant/Refrigerate

Formula: C₂₀H₃₂OSn
Melting Pt: 202…203 °C
MDL Number: MFCD01319066
CAS Number: 148961-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR