5-Amino-2-fluorobenzonitrile ≥98%

Supplier: Matrix Scientific

011106-100G 011106-25G 011106-5G
101780-020EA 392.31 USD
101780-020 101780-016 101780-018
5-Amino-2-fluorobenzonitrile ≥98%
5-Amino-2-fluorobenzonitrile

Hazard: Toxic

Formula: C₇H₅FN₂
MW: 136.13 g/mol
MDL Number: MFCD00277872
CAS Number: 53312-81-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR