Cyclopropylboronic acid pinacol ester ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

009775-5G
101779-996EA 43.08 USD
101779-996
Cyclopropylboronic acid pinacol ester ≥97%
Cyclopropylboronic acid pinacol ester

Hazard: Flammable/Moisture Sensitive/Refrigerate

Formula: C₉H₁₇BO₂
MW: 168.04 g/mol
Boiling Pt: 146 °C
Density: 0.922
Flash Pt: 40 °C
MDL Number: MFCD05663847
CAS Number: 126689-01-8
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Boiling point 146°
Density 0.922

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR