4-Chloro-2-fluorophenylacetonitrile ≥98%
Supplier: Matrix Scientific
Synonyms:
4-Chloro-2-fluorobenzeneacetonitrile
009620-5G
101779-986EA
36.92
USD
101779-986
4-Chloro-2-fluorophenylacetonitrile ≥98%
4-Chloro-2-fluorophenylacetonitrile
Hazard: Toxic
Formula:
C₈H₅ClFN MW: 169.59 g/mol |
MDL Number:
MFCD01312758 CAS Number: 75279-53-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...