4-Chloro-3-fluorobenzoyl chloride
Supplier: Matrix Scientific
009483-5G
009483-25G
101779-968EA
461.54
USD
101779-968
101779-966
4-Chloro-3-fluorobenzoyl chloride
4-Chloro-3-fluorobenzoyl chloride
Hazard: Corrosive/Refrigerate
Formula:
C₇H₃Cl₂FO |
MDL Number:
MFCD02091532 CAS Number: 177787-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...