MATR009204

009204-25G 009204-5G
101779-924EA 64.08 USD
101779-924 101779-922
MATR009204
Purity: 97%
CAS: 383-53-9
MDL Number: MFCD00126489
Molecular Formula: C9H6BrF3O
Molecular Weight: 267,05
Hazard: Corrosive
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR