2,2,2-Trifluoroethyl nonafluorobutanesulfonate ≥97%
Supplier: Matrix Scientific
Synonyms:
2,2,2-Trifluoroethyl 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulphonate
, 2,2,2-Trifluoroethyl Perfluorobutanesulfonate
, 2,2,2-Trifluoroethyl 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonate
, Perfluorobutanesulfonic acid 2,2,2-Trifluoroethyl Ester
, Nonafluorobutanesulfonic acid 2,2,2-Trifluoroethyl Ester
, 2,2,2-Trifluoroethyl Perfluorobutanesulphonate
, Nonafluorobutanesulphonic acid 2,2,2-Trifluoroethyl Ester
, Perfluorobutanesulphonic acid 2,2,2-Trifluoroethyl Ester
Hazard: Toxic/Corrosive
Formula:
C₆H₂F₁₂O₃S MW: 382.12 g/mol Boiling Pt: 138…140 °C |
MDL Number:
MFCD03093638 CAS Number: 79963-95-4 |
Specification Test Results
Boiling point | 138-140° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...