Perfluoro-3,6,9-trioxadecanoic acid ≥98%
Supplier: Matrix Scientific
009137-5G
009137-25G
101779-886EA
315.38
USD
101779-886
101779-884
Perfluoro-3,6,9-trioxadecanoic acid ≥98%
Perfluoro-3,6,9-trioxadecanoic acid
Hazard: Corrosive
Formula:
C₇HF₁₃O₅ Boiling Pt: 170…171 °C Density: 1.8 Flash Pt: >110 °C |
MDL Number:
MFCD00156025 CAS Number: 151772-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...