Perfluoro-3,6-dioxaoctane-1,8-dioic acid ≥98%
Supplier: Matrix Scientific
009099-5G
009099-25G
101779-878EA
200
USD
101779-878
101779-876
Perfluoro-3,6-dioxaoctane-1,8-dioic acid ≥98%
Perfluoro-3,6-dioxaoctane-1,8-dioic acid
Hazard: Corrosive/Irritant
Formula:
C₆H₂F₈O₆ Boiling Pt: 115 °C (0.8 mmHg) |
MDL Number:
MFCD03094537 CAS Number: 55621-21-1 |
Specification Test Results
Boiling point | 115°/0.8mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...