3,3,4,4,4-Pentafluoro-1-butanol ≥97%
Supplier: Matrix Scientific
009075-25G
009075-5G
101779-862EA
607.69
USD
101779-862
101779-860
3,3,4,4,4-Pentafluoro-1-butanol ≥97%
3,3,4,4,4-Pentafluoro-1-butanol
Hazard: Flammable
Formula:
C₄H₅F₅O MW: 164.08 g/mol Boiling Pt: 83…85 °C |
MDL Number:
MFCD03424479 CAS Number: 54949-74-5 |
Specification Test Results
Boiling point | 83-85° |
ND20 | 1.315 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...