Methyl-3-methoxy-2-(trifluoromethyl)propionate ≥97%

Supplier: Matrix Scientific

009031-1G
101779-836EA 235.38 USD
101779-836
Methyl-3-methoxy-2-(trifluoromethyl)propionate ≥97%
Methyl-3-methoxy-2-(trifluoromethyl)propionate

Hazard: Flammable/Irritant

Formula: C₆H₉F₃O₃
MW: 186.13 g/mol
MDL Number: MFCD04038322
CAS Number: 359-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR