Methyl heptafluoroisobutyrate ≥97%

Supplier: Matrix Scientific

Synonyms: Heptafluoroisobutyric Acid Methyl Ester

009028-1G 009028-5G
101779-834EA 184.62 USD
101779-834 101779-832
Methyl heptafluoroisobutyrate ≥97%
Methyl heptafluoroisobutyrate

Purity: 97%
CAS: 680-05-7
MDL Number: MFCD00633403
Molecular Formula: C5H3F7O2
Molecular Weight: 228,07
Boiling Point: 57-58°C
Hazard: Corrosive/Irritant

Formula: C₅H₃F₇O₂
MW: 228.07 g/mol
Boiling Pt: 57…58 °C
MDL Number: MFCD00633403
CAS Number: 680-05-7

Order Now

Specification Test Results

Boiling point 76-77°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR