1,1-Difluoro-1-butene ≥98%
Supplier: Matrix Scientific
008919-10G
101779-794EA
524.62
USD
101779-794
1,1-Difluoro-1-butene ≥98%
1,1-Difluoro-1-butene
Hazard: Flammable Gas
Formula:
C₄H₆F₂ MW: 92.09 g/mol Boiling Pt: 3.7 °C Melting Pt: –144.9 °C Density: 0.9284 |
MDL Number:
MFCD04038010 CAS Number: 407-09-0 |
Specification Test Results
Boiling point | 3.7° |
Density | 1.0216 |
ND20 | 1.3851 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...