1,1-Difluoroacetone ≥97%
Supplier: Matrix Scientific
008915-5G
101779-792EA
74.75
USD
101779-792
1,1-Difluoroacetone ≥97%
1,1-Difluoroacetone
Hazard: Flammable
Specification Test Results
Boiling point | 46.5-46.7° |
Density | 1.1644 |
ND20 | 1.328 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...