N,N-Diethyl-2,2,2-trifluoroacetamide ≥99%

Supplier: Matrix Scientific

008913-5G
101779-788EA 66.15 USD
101779-788
N,N-Diethyl-2,2,2-trifluoroacetamide ≥99%
N,N-Diethyl-2,2,2-trifluoroacetamide

Hazard: Flammable/Irritant

Formula: C₆H₁₀F₃NO
MW: 169.15 g/mol
Boiling Pt: 65…67 °C (24 mmHg)
Density: 1.142
Flash Pt: 43 °C
MDL Number: MFCD00192034
CAS Number: 360-92-9

Order Now

Specification Test Results

Boiling point 65-67°/24m
Density 1.2
ND20 1.38

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR