Chlorofluoroacetic acid ≥97%
Supplier: Matrix Scientific
008872-5G
008872-1G
101779-786EA
607.69
USD
101779-786
101779-784
Chlorofluoroacetic acid ≥97%
Chlorofluoroacetic acid
Hazard: Corrosive/Irritant
Formula:
C₂H₂ClFO₂ MW: 112.49 g/mol Boiling Pt: 162 °C Density: 1.532 |
MDL Number:
MFCD00871521 CAS Number: 471-44-3 |
Specification Test Results
Boiling point | 162° |
Density | 1.532 |
ND20 | 1.4105 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...