2-Bromopentafluoropropene ≥97%

Supplier: Matrix Scientific

008846-5G
101779-782EA 283.08 USD
101779-782
2-Bromopentafluoropropene ≥97%
2-Bromopentafluoropropene

Hazard: Toxic/Irritant

Formula: C₃BrF₅
Boiling Pt: 26…28 °C
MDL Number: MFCD03094565
CAS Number: 431-49-2

Order Now

Specification Test Results

Boiling point 25-26°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR