Bromochlorodifluoromethane ≥97%

Supplier: Matrix Scientific

008821-25G 008821-100G
101779-768EA 946.15 USD
101779-768 101779-770
Bromochlorodifluoromethane ≥97%
Bromochlorodifluoromethane

Hazard: Ozone Depleter/Nonflammable Gas

Formula: BrClCF₂
Boiling Pt: –2.5 °C
Melting Pt: –160.5 °C
Density: 1.917
CAS Number: 353-59-3
UN: 1956
ADR: 2,

Order Now

Specification Test Results

Boiling point -2.5°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR