Allyl-2,2,3,3-tetrafluoropropyl ether ≥97%
Supplier: Matrix Scientific
008794-25G
008794-5G
101779-762EA
267.69
USD
101779-762
101779-760
Allyl-2,2,3,3-tetrafluoropropyl ether ≥97%
Allyl-2,2,3,3-tetrafluoropropyl ether
Hazard: Flammable/Irritant
Formula:
C₆H₈F₄O MW: 172.12 g/mol Boiling Pt: 110 °C (740 mmHg) Density: 1.18 |
MDL Number:
MFCD02093319 CAS Number: 681-68-5 |
Specification Test Results
Boiling point | 110°/740mm |
Density | |
ND20 | 1.3471 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...