2-Picolyl chloride hydrochloride ≥98%
Supplier: Matrix Scientific
Synonyms:
2-(Chloromethyl)pyridine hydrochloride
008729-100G
101779-732EA
75.38
USD
101779-732
2-Picolyl chloride hydrochloride ≥98%
2-Picolyl chloride hydrochloride
Hazard: Corrosive/Hygroscopic
Formula:
C₆H₇Cl₂N MW: 164.03 g/mol Melting Pt: 120…124 °C |
MDL Number:
MFCD00012811 CAS Number: 6959-47-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...