2-Amino-3,6-difluoro-4-(trifluoromethyl)pyridine ≥96%

Supplier: Matrix Scientific

008714-250MG
101779-722EA 129.23 USD
101779-722
2-Amino-3,6-difluoro-4-(trifluoromethyl)pyridine ≥96%
2-Amino-3,6-difluoro-4-(trifluoromethyl)pyridine

Hazard: Toxic

Formula: C₆H₃F₅N₂
Melting Pt: 66…68 °C
MDL Number: MFCD03840200
CAS Number: 675602-89-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 66-68°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR