2,2-Dichloro-1,1,1-trifluoroethane ≥99%

Supplier: Matrix Scientific

007656-100G
101779-694EA 369.23 USD
101779-694
2,2-Dichloro-1,1,1-trifluoroethane ≥99%
2,2-Dichloro-1,1,1-trifluoroethane

TCA listed: T
Hazard: Ozone Depleter/Irritant/Nonflammable Gas/Store in Freezer

Formula: Cl₂HCCF₃
Boiling Pt: 27.1 °C
Melting Pt: –107 °C
Density: 1.475
CAS Number: 306-83-2
UN: 1230
ADR: 3,II

Order Now

Specification Test Results

Boiling point 27.1°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR