N-Methylhexylamine ≥97%
Supplier: Matrix Scientific
Synonyms:
N-Methylhexan-1-amine
007627-5G
101779-680EA
144.62
USD
101779-680
N-Methylhexylamine ≥97%
N-Methylhexylamine
Hazard: Flammable/Corrosive
Formula:
C₇H₁₇N MW: 115.22 g/mol Boiling Pt: 140…142 °C Density: 0.76 Flash Pt: 23 °C |
MDL Number:
MFCD00042908 CAS Number: 35161-70-7 |
Specification Test Results
Boiling point | 140°-142° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...