N,N'-Di-n-propylethylenediamine ≥97%
Supplier: Matrix Scientific
Synonyms:
N1,N1-Dipropylethane-1,2-diamine
, N,N'-Di(n-propyl)ethylenediamine
007584-5G
007584-25G
101779-654EA
197.06
USD
101779-654
101779-652
N,N'-Di-n-propylethylenediamine ≥97%
N,N'-Di-n-propylethylenediamine
Hazard: Corrosive
Formula:
C₈H₂₀N₂ MW: 144.262 g/mol Boiling Pt: 65 °C (2 mmHg) |
MDL Number:
MFCD00015199 CAS Number: 17361-75-0 |
Specification Test Results
Boiling point | 65°/2mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...