2,6-Dimethylpiperazine ≥97%
Supplier: Matrix Scientific
007579-100G
007579-25G
007579-5G
101779-648EA
255.38
USD
101779-648
101779-646
101779-650
2,6-Dimethylpiperazine ≥97%
2,6-Dimethylpiperazine
Hazard: Flammable Solid
Formula:
C₆H₁₄N₂ Boiling Pt: 162 °C Melting Pt: 108…111 °C Flash Pt: 45 °C |
MDL Number:
MFCD00005956 CAS Number: 108-49-6 UN: 1325 ADR: 4.1,II |
Specification Test Results
Boiling point | 162° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...