N,N-Dimethyl-tert-butylamine ≥97%

Supplier: Matrix Scientific

007578-25G
101779-644EA 140 USD
101779-644
N,N-Dimethyl-tert-butylamine ≥97%
N,N-Dimethyl-tert-butylamine

Hazard: Corrosive

Formula: C₆H₁₅N
Boiling Pt: 88…90 °C
MDL Number: MFCD03452796
CAS Number: 918-02-5

Order Now

Specification Test Results

Boiling point 88°-90°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR