(Dimethylamino)acetone ≥97%

Supplier: Matrix Scientific

007577-5G
101779-642EA 576.92 USD
101779-642
(Dimethylamino)acetone ≥97%
(Dimethylamino)acetone

Hazard: Flammable/Light Sensitive/Air Sensitive

Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 119 °C
Density: 0.883
Flash Pt: 30 °C
MDL Number: MFCD00008766
CAS Number: 15364-56-4
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Boiling point 119°
Density 0.883
ND20 1.414

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR