(Diethylamino)acetone ≥97%

Supplier: Matrix Scientific

Synonyms: Diethylaminoacetone

007557-25G 007557-5G
101779-612EA 792.31 USD
101779-612 101779-610
(Diethylamino)acetone ≥97%
(Diethylamino)acetone

Hazard: Flammable/Irritant

Formula: C₇H₁₅NO
MW: 129.2 g/mol
Boiling Pt: 64 °C ( 16 mmHg)
MDL Number: MFCD00009050
CAS Number: 1620-14-0
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Boiling point 64°/ 16mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR