2,6-Dichlorobenzylamine ≥97%

Supplier: Matrix Scientific

Synonyms: (2,6-Dichlorophenyl)methanamine

007554-1G 007554-5G 007554-25G
101779-606EA 90.77 USD
101779-606 101779-604 101779-608
2,6-Dichlorobenzylamine ≥97%
2,6-Dichlorobenzylamine

Hazard: Corrosive/Air Sensitive

Formula: C₇H₇Cl₂N
MW: 176.04 g/mol
Boiling Pt: 108 °C ( 8 mmHg)
Melting Pt: 40 °C
MDL Number: MFCD00047928
CAS Number: 6575-27-5
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Boiling point 108°/ 8mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR