N,N'-Di-n-butylethylenediamine ≥97%

Supplier: Matrix Scientific

007552-25G
101779-602EA 121.54 USD
101779-602
N,N'-Di-n-butylethylenediamine ≥97%
N,N'-Di-n-butylethylenediamine

TCA listed: T
Hazard: Corrosive

Formula: C₁₀H₂₄N₂
Boiling Pt: 92 °C ( 3 mmHg)
MDL Number: MFCD00015230
CAS Number: 4013-95-0

Order Now

Specification Test Results

Boiling point 92°/ 3mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR